Thermo Scientific Chemicals

Isobutyl methacrylate, 99.5+%, stab. with 10ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: 041948.K2
1 L, Each
Thermo Scientific Chemicals

Isobutyl methacrylate, 99.5+%, stab. with 10ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: 041948.K2
1 L, Each
Quantity
Catalog number: 041948.K2
also known as 041948-K2
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
97-86-9
IUPAC Name
2-methylpropyl 2-methylprop-2-enoate
Molecular Formula
C8H14O2
InChI Key
RUMACXVDVNRZJZ-UHFFFAOYSA-N
SMILES
CC(C)COC(=O)C(C)=C
Appearance (Color)
Colorless to very pale yellow
Form
Liquid
Stabilizer
4-methoxyphenol: 9 to 11 ppm
Assay (GC)
≥99.5%

Description

Plastic industry: used in the manufacture of containing active hydroxyl acrylic resin. Coatings with epoxy resin, diisocyanate and melamine formaldehyde resin configuration used for preparing a two-component coating. In oil industry as additives for lubricating oil washing. In electronics industry used as electron microscope to take off the water. Textile industry used in the manufacture of adhesive fabric. Used as chemical reagents in analytical chemistry. In addition, also used for water immiscible embedding agent, used in the synthesis of medical polymer materials, thermosetting coatings and adhesives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text