Thermo Scientific Chemicals
3-(Benzylamino)propionitrile, 97+%, Thermo Scientific Chemicals
Catalog number: 041949.22
100 g, Each
Thermo Scientific Chemicals
3-(Benzylamino)propionitrile, 97+%, Thermo Scientific Chemicals
Catalog number: 041949.22
100 g, Each
Quantity
Catalog number: 041949.22
also known as 041949-22
Price (USD)
Quantity
-
Have Questions?
Description
3-(Benzyl amino)propionitrile was used as starting reagent in the synthesis of (3R,4S)-1-benzyl-4-phenylpyrrolidine-3-carboxylic acid, 3-[benzyl-(3,5-dimethyl-pyrazol-1-ylmethyl)-amino]-propionitrile. It undergoes aza-type Michael reaction with 1-hydroxymethyl-5-methyl-1H-pyrazole-3-carboxylic acid ethyl ester under mild conditions