Thermo Scientific Chemicals

Magnesium citrate, dibasic hydrate, Thermo Scientific Chemicals

Catalog number: 041954.30
250 g, Each
Thermo Scientific Chemicals

Magnesium citrate, dibasic hydrate, Thermo Scientific Chemicals

Catalog number: 041954.30
250 g, Each
Quantity
Catalog number: 041954.30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
119851-23-9
IUPAC Name
magnesium(2+) 2-(carboxymethyl)-2-hydroxybutanedioate
Molecular Formula
C6H6MgO7
InChI Key
DIXGJWCZQHXZNR-UHFFFAOYNA-L
SMILES
[Mg++].OC(=O)CC(O)(CC([O-])=O)C([O-])=O
Form
Powder
Assay from Supplier's CofA
Mg: 7.5-10.5%
Water Content
Waters of hydration: 1-6

Description

Magnesium citrate is used to clean stool from the intestines before surgery. It is also used as a laxative for relief of constipation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text