Magnesium citrate, dibasic hydrate, Thermo Scientific Chemicals
Magnesium citrate, dibasic hydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Magnesium citrate, dibasic hydrate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number 041954.A1
also known as 041954-A1
Price (USD)/ Each
270.65
Online exclusive
301.00 
Save 30.35 (10%)
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
270.65
Online exclusive
301.00 
Save 30.35 (10%)
Add to cart
Magnesium citrate, dibasic hydrate, Thermo Scientific Chemicals
Catalog number041954.A1
Price (USD)/ Each
270.65
Online exclusive
301.00 
Save 30.35 (10%)
-
Add to cart
Chemical Identifiers
CAS119851-23-9
IUPAC Namemagnesium(2+) 2-(carboxymethyl)-2-hydroxybutanedioate
Molecular FormulaC6H6MgO7
InChI KeyDIXGJWCZQHXZNR-UHFFFAOYNA-L
SMILES[Mg++].OC(=O)CC(O)(CC([O-])=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay from Supplier's CofAMg: 7.5-10.5%
Water ContentWaters of hydration: 1-6
Magnesium citrate is used to clean stool from the intestines before surgery. It is also used as a laxative for relief of constipation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only