Thermo Scientific Chemicals

Lead(II) tetrafluoroborate, 50% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: 041956.A1
1 kg, Each
Thermo Scientific Chemicals

Lead(II) tetrafluoroborate, 50% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: 041956.A1
1 kg, Each
Quantity
Catalog number: 041956.A1
also known as 041956-A1
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13814-96-5
IUPAC Name
λ²-lead(2+) bis(tetrafluoroboranuide)
Molecular Formula
B2F8Pb
InChI Key
UQGXAENNKXXYNF-UHFFFAOYSA-N
SMILES
[Pb++].F[B-](F)(F)F.F[B-](F)(F)F
Concentration (weight/weight)
50% min

Description

Used in tin-lead electroplatingLead(II) tetrafluoroborate is used as a catalyst involved in calixarene formation through hydrogen-bonded dimmers and in the preparation of linear polyesters. It is involved in the development and optimization of a bipolar lead-boron tetrafluoride accumulator. It is an electroplating solution for coating metal objects with lead and curing agent for epoxy resins. Further, it is used in printing lines, tin-lead alloy plating, lead soldering and also as a circuit board tin-lead alloy plating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text