Thermo Scientific Chemicals

Potassium tellurite monohydrate, 97%, Thermo Scientific Chemicals

Catalog number: 041959.14
25 g, Each
Thermo Scientific Chemicals

Potassium tellurite monohydrate, 97%, Thermo Scientific Chemicals

Catalog number: 041959.14
25 g, Each
Quantity
Catalog number: 041959.14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
123333-66-4
Assay (unspecified)
96.1% min

Description

Potassium tellurite, in association with agar is used as a selective medium for growth of clauberg (bacteria). It is used for the selective isolation of staphylococci and corynebacteria, which is used to reduce TeO3 2- to tellurium element.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text