Thermo Scientific Chemicals

Indium(III) chloride, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 041977.30
250 g, Each
Thermo Scientific Chemicals

Indium(III) chloride, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 041977.30
250 g, Each
Quantity
Catalog number: 041977.30
also known as 041977-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10025-82-8
IUPAC Name
indium(3+) trichloride
Molecular Formula
Cl3In
InChI Key
PSCMQHVBLHHWTO-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[In+3]
Appearance (Color)
White to pale cream
Form
Crystalline powder or powder and/or chunks
Total Metal Impurities
!<0.01%

Description

Indium(III) chloride is used in organic reactions such as Friedel-crafts acylation and Diels-Alder reaction as a Lewis acid catalyst. It is an efficient catalyst for thioacetalization and its chemoselectivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text