Thermo Scientific Chemicals

Antimony(V) chloride, 98%, Thermo Scientific Chemicals

Catalog number: 041978.18
50 g, Each
Thermo Scientific Chemicals

Antimony(V) chloride, 98%, Thermo Scientific Chemicals

Catalog number: 041978.18
50 g, Each
Quantity
Catalog number: 041978.18
also known as 041978-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7647-18-9
IUPAC Name
antimony pentachloride
Molecular Formula
Cl5Sb
InChI Key
VMPVEPPRYRXYNP-UHFFFAOYSA-I
SMILES
Cl[Sb](Cl)(Cl)(Cl)Cl
Assay (unspecified)
≥97.5%

Description

Antimony(V) chloride is used as a catalyst and an analytical reagent for testing alkaloids and cesium. It is also used as a polymerization catalyst as well as involved in the chlorination of organic compounds. It acts as a Lewis acid and a strong oxidizing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text