Potassium hydrogen iodate, primary standard, ACS, 99.95-100.05%, Thermo Scientific Chemicals
Potassium hydrogen iodate, primary standard, ACS, 99.95-100.05%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium hydrogen iodate, primary standard, ACS, 99.95-100.05%, Thermo Scientific Chemicals

Maximum level of impurities: Insoluble matter 0.01%, SO4 0.005%, Fe 0.001%, Cl 0.02%, Heavy Metals (as Pb) 0.001%
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 041986.14
also known as 041986-14
Price (USD)/ Each
61.65
Online exclusive
68.70 
Save 7.05 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
61.65
Online exclusive
68.70 
Save 7.05 (10%)
Add to cart
Potassium hydrogen iodate, primary standard, ACS, 99.95-100.05%, Thermo Scientific Chemicals
Catalog number041986.14
Price (USD)/ Each
61.65
Online exclusive
68.70 
Save 7.05 (10%)
-
Add to cart
Chemical Identifiers
CAS13455-24-8
IUPAC Namepotassium iodic acid iodate
Molecular FormulaHI2KO6
InChI KeyACAYDTMSDROWHW-UHFFFAOYSA-M
SMILES[K+].O[I](=O)=O.[O-][I](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.95-100.05% KH(IO₃)₂
Impurity contentCl 0.02%
Impurity contentFe 0.001%
Impurity contentSO₄ 0.005%
Insoluble material0.01%
View more
Potassium hydrogen iodate acts as an oxidizing agent. Further, it serves as an intermediate for the synthesis of potassium iodate and also used as an analysis reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only