Europium(III) phosphate hydrate, 99.9% (REO), Thermo Scientific Chemicals
Europium(III) phosphate hydrate, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Europium(III) phosphate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 041989.06
also known as 041989-06
Price (USD)/ Each
166.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
166.00
Add to cart
Europium(III) phosphate hydrate, 99.9% (REO), Thermo Scientific Chemicals
Catalog number041989.06
Price (USD)/ Each
166.00
-
Add to cart
Chemical Identifiers
CAS14913-20-3
IUPAC Nameeuropium(3+) phosphate
Molecular FormulaEuO4P
InChI KeyKJNZTHUWRRLWOA-UHFFFAOYSA-K
SMILES[Eu+3].[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Europium finds application in optical coating and thin film application.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only