Lanthanum(III) phosphate hydrate, 99.99% (REO), Thermo Scientific Chemicals
Lanthanum(III) phosphate hydrate, 99.99% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lanthanum(III) phosphate hydrate, 99.99% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 041990.06
also known as 041990-06
Price (USD)/ Each
50.20
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
50.20
Add to cart
Lanthanum(III) phosphate hydrate, 99.99% (REO), Thermo Scientific Chemicals
Catalog number041990.06
Price (USD)/ Each
50.20
-
Add to cart
Chemical Identifiers
CAS14913-14-5
IUPAC Namelanthanum(3+) phosphate
Molecular FormulaLaO4P
InChI KeyLQFNMFDUAPEJRY-UHFFFAOYSA-K
SMILES[La+3].[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.01% max.
It is an active pharmacetical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only