Yttrium(III) phosphate hydrate, 99.99% (REO), Thermo Scientific Chemicals
Yttrium(III) phosphate hydrate, 99.99% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Yttrium(III) phosphate hydrate, 99.99% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
Catalog number 041991.09
also known as 041991-09
Price (USD)/ Each
153.65
Online exclusive
171.00 
Save 17.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
153.65
Online exclusive
171.00 
Save 17.35 (10%)
Add to cart
Yttrium(III) phosphate hydrate, 99.99% (REO), Thermo Scientific Chemicals
Catalog number041991.09
Price (USD)/ Each
153.65
Online exclusive
171.00 
Save 17.35 (10%)
-
Add to cart
Chemical Identifiers
CAS34054-55-2
IUPAC Nameyttrium(3+) phosphate
Molecular FormulaO4PY
InChI KeyUXBZSSBXGPYSIL-UHFFFAOYSA-K
SMILES[Y+3].[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.01% max.
The most important use of yttrium is in making phosphors, particularly the red phosphors used in television set cathode ray tube (CRT) displays and in LEDs. It is also used in the production of electrodes, electrolytes, electronic filters, lasers and superconductors; various medical applications; and the tracing of various materials to enhance their properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only