Thermo Scientific™

Fullerene powder, 99.9+% C{60}, Thermo Scientific Chemicals

Catalog number: 042007.MD
250 mg, Each
Thermo Scientific™

Fullerene powder, 99.9+% C{60}, Thermo Scientific Chemicals

Catalog number: 042007.MD
250 mg, Each
Quantity
Catalog number: 042007.MD
also known as 042007-MD
Price (USD)
Have Questions?

Chemical Identifiers

CAS
99685-96-8
IUPAC Name
(C60-Ih)[5,6]fullerene
Molecular Formula
C60
InChI Key
XMWRBQBLMFGWIX-UHFFFAOYSA-N
SMILES
C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41
Form
Powder
Purity
≥99.9% C60

Description

Fullerene powder is useful for biomedical applications such as the design of high-performance MRI contrast agents, x-ray imaging contrast agents and photodynamic therapy. It is also essential for single-walled carbon nanotubes with narrow diameter-distribution. It is used as light-activated antimicrobial agent. It also finds application in the field of nanotechnology, heat resistance and superconductivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text