Thermo Scientific Chemicals
Stainless Steel gauze, 150 mesh woven from 0.066mm (0.0026in) dia wire, Type 304, Thermo Scientific Chemicals
Catalog number: 042010.RU
30 x 60 cm, Each
Thermo Scientific Chemicals
Stainless Steel gauze, 150 mesh woven from 0.066mm (0.0026in) dia wire, Type 304, Thermo Scientific Chemicals
Catalog number: 042010.RU
30 x 60 cm, Each
Quantity
Catalog number: 042010.RU
also known as 042010-RU