Stainless Steel gauze, 400 mesh woven from 0.028mm (0.0011in) dia wire, twilled weave, Type 304, Thermo Scientific Chemicals
Stainless Steel gauze, 400 mesh woven from 0.028mm (0.0011in) dia wire, twilled weave, Type 304, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Stainless Steel gauze, 400 mesh woven from 0.028mm (0.0011in) dia wire, twilled weave, Type 304, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
15 x 30 cm
15 x 15 cm
Catalog number 042012.CF
also known as 042012-CF
Price (USD)/ Each
226.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
266.00 
Save 39.35 (15%)
-
Add to cart
Quantity:
15 x 30 cm
Request bulk or custom format
Price (USD)/ Each
226.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
266.00 
Save 39.35 (15%)
Add to cart
Stainless Steel gauze, 400 mesh woven from 0.028mm (0.0011in) dia wire, twilled weave, Type 304, Thermo Scientific Chemicals
Catalog number042012.CF
Price (USD)/ Each
226.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
266.00 
Save 39.35 (15%)
-
Add to cart
Specifications
Chemical Name or MaterialStainless Steel gauze, Type 304
Name NoteWoven from 0.028mm (0.0011 in.) dia. wire
Health Hazard 1H317-H350-H372
Health Hazard 2GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
View more
It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only