n-Butyl dodecanoate, 99+%, Thermo Scientific Chemicals
n-Butyl dodecanoate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Butyl dodecanoate, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
500 mL
Catalog number 042024.AE
also known as 042024-AE
Price (USD)/ Each
51.80
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
51.80
Add to cart
n-Butyl dodecanoate, 99+%, Thermo Scientific Chemicals
Catalog number042024.AE
Price (USD)/ Each
51.80
-
Add to cart
Specifications
Chemical Name or Materialn-Butyl dodecanoate
CAS106-18-3
Melting Point-10°C
Recommended StorageAmbient temperatures
Density0.86 g/mL
View more
n-Butyl dodecanoate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only