Aluminum spheres, 2.38mm (0.094in) dia, 99% (metals basis), Thermo Scientific Chemicals
Aluminum spheres, 2.38mm (0.094in) dia, 99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum spheres, 2.38mm (0.094in) dia, 99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 Pc.
25 Pc.
Catalog number 042029.KN
also known as 042029-KN
Price (USD)/ Each
127.00
-
Add to cart
Quantity:
100 Pc.
Request bulk or custom format
Price (USD)/ Each
127.00
Add to cart
Aluminum spheres, 2.38mm (0.094in) dia, 99% (metals basis), Thermo Scientific Chemicals
Catalog number042029.KN
Price (USD)/ Each
127.00
-
Add to cart
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only