Thermo Scientific Chemicals

Dodecanoic acid, typically 99.5%, Thermo Scientific Chemicals

Catalog number: 042038.A1
1 kg, Each
Thermo Scientific Chemicals

Dodecanoic acid, typically 99.5%, Thermo Scientific Chemicals

Catalog number: 042038.A1
1 kg, Each
Quantity
Catalog number: 042038.A1
also known as 042038-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
143-07-7
IUPAC Name
dodecanoic acid
Molecular Formula
C12H24O2
InChI Key
POULHZVOKOAJMA-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCC(O)=O
Appearance (Color)
White
Assay from Supplier's CofA
≥99.4% (GC)
Form
Crystals or flakes or waxy or fused solid

Description

Dodecanoic acid is used in the preparation of cosmetics, soaps, alkyd resins and wetting agents. It is also used to measure the molar mass of an unknown substance through freezing point depression. It is also used as a food additive and an active component in a treatment for acne. In addition to this, it is a substrate for acylation of certain proteins based on the murine studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text