Thermo Scientific Chemicals

Gallium(III) 2,4-pentanedionate, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 042086.06
5 g, Each
Thermo Scientific Chemicals

Gallium(III) 2,4-pentanedionate, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 042086.06
5 g, Each
Quantity
Catalog number: 042086.06
also known as 042086-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14405-43-7
IUPAC Name
gallium(3+) tris(2,4-dioxopentan-3-ide)
Molecular Formula
C15H21GaO6
InChI Key
IMMRYVXLCAMRNK-UHFFFAOYSA-N
SMILES
[Ga+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Total Metal Impurities
0.01% max.

Description

It is used in research on Ga-containing materials. Gallium oxide thin films can be created with atomic layer epitaxy (ALE) by combining gallium acetylacetonate with either water or ozone as the precursor. It can also be used for the synthesis of high purity gallium nitride nano-wires and nano-needles at low temperatures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text