Thermo Scientific Chemicals
Indium(III) isopropoxide, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number: 042098.09
10 g, Each
Thermo Scientific Chemicals
Indium(III) isopropoxide, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number: 042098.09
10 g, Each
Quantity
Catalog number: 042098.09
also known as 042098-09
Price (USD)
Quantity
-
Have Questions?
Description
Indium(III) isopropoxide acts as a precursor for the preparation of indium oxide containing films. It finds application in nano-materials and thin films display. Furthermore, it is used in imaging catalysis and in preparation of sensors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under argon