Vanadium slug, 11mm (0.43in) dia x 4mm (0.16in) thick, 99.8% (metals basis), Thermo Scientific Chemicals
Vanadium slug, 11mm (0.43in) dia x 4mm (0.16in) thick, 99.8% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Vanadium slug, 11mm (0.43in) dia x 4mm (0.16in) thick, 99.8% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 Pc.
5 Pc.
Catalog number 042190.KM
also known as 042190-KM
Price (USD)/ Each
567.65
Online exclusive
631.00 
Save 63.35 (10%)
-
Add to cart
Quantity:
25 Pc.
Request bulk or custom format
Price (USD)/ Each
567.65
Online exclusive
631.00 
Save 63.35 (10%)
Add to cart
Vanadium slug, 11mm (0.43in) dia x 4mm (0.16in) thick, 99.8% (metals basis), Thermo Scientific Chemicals
Catalog number042190.KM
Price (USD)/ Each
567.65
Online exclusive
631.00 
Save 63.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-62-2
IUPAC Namevanadium
Molecular FormulaV
InChI KeyLEONUFNNVUYDNQ-UHFFFAOYSA-N
SMILES[V]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.8% (metals basis)
Thickness4mm (0.16in) thick.
FormSlug, 11mm (0.43in) diameter.
Appearance (Color)White silver
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only