Thermo Scientific Chemicals

Indium oxide, tin doped, polymeric precursor, Oxide ≈20 wt%, Thermo Scientific Chemicals

Catalog number: 042229.09
10 g, Each
Thermo Scientific Chemicals

Indium oxide, tin doped, polymeric precursor, Oxide ≈20 wt%, Thermo Scientific Chemicals

Catalog number: 042229.09
10 g, Each
Quantity
Catalog number: 042229.09
also known as 042229-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1312-43-2
IUPAC Name
diindium(3+) trioxidandiide
Molecular Formula
In2O3
InChI Key
PJXISJQVUVHSOJ-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[In+3].[In+3]
Form
viscous liquid
Appearance (Color)
Yellow to brown
Assay (unspecified)
>99% ( metals basis)

Description

It is used in the preparation of transparent electrical conducting electrodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text