Thermo Scientific Chemicals
Hexavalent Chromium, standard solution, Specpure™ Cr∣+6 1000μg/mL, Thermo Scientific Chemicals
Catalog number: 042234.AP
500 mL, Each
Thermo Scientific Chemicals
Hexavalent Chromium, standard solution, Specpure™ Cr∣+6 1000μg/mL, Thermo Scientific Chemicals
Catalog number: 042234.AP
500 mL, Each
Quantity
Catalog number: 042234.AP
also known as 042234-AP
Price (USD)
Quantity
-
Have Questions?
Starting Material
Na2CrO4
Concentration
1,000?g/mL ?1%
Description
Hexavalent Chromium, standard solution is used in ion chromatography for measurement and determination of soluble Cr(VI).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov