Thermo Scientific Chemicals

Polyethylene oxide, M.W. 100,000, Thermo Scientific Chemicals

Catalog number: 042236.22
100 g, Each
Thermo Scientific Chemicals

Polyethylene oxide, M.W. 100,000, Thermo Scientific Chemicals

Catalog number: 042236.22
100 g, Each
Quantity
Catalog number: 042236.22
also known as 042236-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25322-68-3
Molecular Formula
(C2H4O)n
InChI Key
LYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES
[H]OCCO
Appearance (Color)
White to off-white
Assay (unspecified)
Viscosity: 30-50 mPa sec
Comment
Molecular Weight: 100,000 (approx.)
Form
Powder

Description

Polyethylene oxide is used in conjunction with carbon black to form a conductive composite. It serves as thermoplastic material that is readily calendared, extruded, injection molded or cast, to provide even greater formulation and processing flexibility. It also provides binding, thickening, lubricity, water retention, and film formation benefits to deliver excellent performance in various applications viz. pharmaceuticals, mining, paper and cleaning products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text