Thermo Scientific Chemicals

Polyethylene oxide, M.W. 300,000, Thermo Scientific Chemicals

Catalog number: 042237.22
100 g, Each
Thermo Scientific Chemicals

Polyethylene oxide, M.W. 300,000, Thermo Scientific Chemicals

Catalog number: 042237.22
100 g, Each
Quantity
Catalog number: 042237.22
also known as 042237-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25322-68-3
Molecular Formula
(C2H4O)n
InChI Key
LYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES
[H]OCCO
Form
Powder
Comment
Purchased in U.K. and U.S.
Assay from Suppliers CofA
Volatiles: ≤0.5% (U.S. specification)
Appearance (Color)
White to pale cream
Assay from Suppliers CofA
Viscosity: 600-1200cPs (c=5%, water, 25°C) (U.K. specification)

Description

Polyethylene oxide, M.W. 300,000 is used to impart viscosity to and modify flow of aqueous solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text