Amberlite™ IRA-200C(Na), ion exchange resin
Amberlite™ IRA-200C(Na), ion exchange resin
Thermo Scientific Chemicals

Amberlite™ IRA-200C(Na), ion exchange resin

Styrene-DVB; Strongly acidic macroreticular cation exchange resin | Styrene-DVB
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Catalog number 042252.30
also known as 042252-30
Price (USD)
30.50
Each
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
30.50
Each
Add to cart
Specifications
Chemical Name or MaterialAmberlite IRA-200C(Na)
MDL NumberMFCD01864001
Molecular FormulaStyrene-DVB
OdorOdorless
Physical FormBeads
View more
Ask our AI about this Product
Amberlite™ IRA-200C(Na), ion exchange resin is used for makeup demineralization, hot process softeners and sodium cycle condensate polishers. Further, it is used in pharmaceuticals and food processing. In addition to this, it is used in portable water applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link