Thermo Scientific Chemicals
Amberlite™ IRA-200C(Na), ion exchange resin, Thermo Scientific Chemicals
Catalog number: 042252.A1
1 kg, Each
Thermo Scientific Chemicals
Amberlite™ IRA-200C(Na), ion exchange resin, Thermo Scientific Chemicals
Catalog number: 042252.A1
1 kg, Each
Quantity
Catalog number: 042252.A1
also known as 042252-A1
Price (USD)
Quantity
-
Have Questions?
Form
Spherical particle
Harmonic mean size
0.600 to 0.850 mm
Appearance (Color)
Pale yellow to pale brown
Exchange capacity
> 1.70 eq/liter
Description
Amberlite™ IRA-200C(Na), ion exchange resin is used for makeup demineralization, hot process softeners and sodium cycle condensate polishers. Further, it is used in pharmaceuticals and food processing. In addition to this, it is used in portable water applications.