Thermo Scientific Chemicals

Amberlite™ IRA-67, ion exchange resin, Thermo Scientific Chemicals

Catalog number: 042253.30
250 g, Each
Thermo Scientific Chemicals

Amberlite™ IRA-67, ion exchange resin, Thermo Scientific Chemicals

Catalog number: 042253.30
250 g, Each
Quantity
Catalog number: 042253.30
also known as 042253-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
80747-90-6
Particle Size
<300 µm: ≤1.0%
Particle Size
>1180 µm: ≤5.0%
Particle Size
Particle Diameter: 500-750 µm
Assay (special)
Uniformity Coefficient: ≤1.80
Assay (special)
Total Exchange Capacity: 1.60-1.71 eq/L

Description

Amberlite™ IRA-67 is used for de-acidification and desalination of water and for isolation of acidic natural products. It allows for effective adsorption and desorption of naturally occurring organic molecules such as humic and fulvic acids that are present in many water supplies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text