Stainless Steel Foil, 0.2mm (0.008 in.) thick, Thermo Scientific Chemicals
Stainless Steel Foil, 0.2mm (0.008 in.) thick, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Stainless Steel Foil, 0.2mm (0.008 in.) thick, Thermo Scientific Chemicals

Have Questions?
Quantity:
100 x 100 mm
Catalog number 042255.GH
also known as 042255-GH
Price (USD)/ Each
46.60
-
Add to cart
Quantity:
100 x 100 mm
Request bulk or custom format
Price (USD)/ Each
46.60
Add to cart
Stainless Steel Foil, 0.2mm (0.008 in.) thick, Thermo Scientific Chemicals
Catalog number042255.GH
Price (USD)/ Each
46.60
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialStainless Steel
ColorSilver
Name NoteType 304
Health Hazard 1H317-H350-H372
Health Hazard 3P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
View more
It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only