Thermo Scientific Chemicals

Acetonitrile-d{3}, 100% (Isotopic), Thermo Scientific Chemicals

Catalog number: 042266.SA
2 Ea., Each
Thermo Scientific Chemicals

Acetonitrile-d{3}, 100% (Isotopic), Thermo Scientific Chemicals

Catalog number: 042266.SA
2 Ea., Each
Quantity
Catalog number: 042266.SA
also known as 042266-SA
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2206-26-0
IUPAC Name
(²H₃)acetonitrile
Molecular Formula
C2H3N
InChI Key
WEVYAHXRMPXWCK-FIBGUPNXSA-N
SMILES
[2H]C([2H])([2H])C#N
Assay (unspecified)
99.96% min., isotopic enrichment

Description

Isotope labeled Acetonitrile is used as a polar aprotic solvent in organic synthesis. Acetonitrile-d3 is used as IR and NMR solvent. Probe for zeolite acidity. Cartridge eluent in HPLC- SPE (solid-phase extraction)-NMR technique.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text