Bromobenzene-d{5}, 99% (Isotopic), Thermo Scientific Chemicals
Bromobenzene-d{5}, 99% (Isotopic), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bromobenzene-d{5}, 99% (Isotopic), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 042269.09
also known as 042269-09
Price (USD)/ Each
235.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
277.00 
Save 41.35 (15%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
235.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
277.00 
Save 41.35 (15%)
Add to cart
Bromobenzene-d{5}, 99% (Isotopic), Thermo Scientific Chemicals
Catalog number042269.09
Price (USD)/ Each
235.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
277.00 
Save 41.35 (15%)
-
Add to cart
Specifications
Chemical Name or MaterialBromobenzene-d5
Density1.55 g/mL
Flash Point51°C (124°F)
OdorAromatic
Physical FormLiquid
View more
The labeled version of bromobenzene , used as a general reagent in palladium-catalyzed reaction and in the synthesize of Grignard reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only