Thermo Scientific Chemicals

Cyclohexane-d{12}, 99.5%(Isotopic), Thermo Scientific Chemicals

Catalog number: 042271.KV
10 Ea., Each
Thermo Scientific Chemicals

Cyclohexane-d{12}, 99.5%(Isotopic), Thermo Scientific Chemicals

Catalog number: 042271.KV
10 Ea., Each
Quantity
Catalog number: 042271.KV
also known as 042271-KV
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1735-17-7
IUPAC Name
(²H₁₂)cyclohexane
Molecular Formula
C6H12
InChI Key
XDTMQSROBMDMFD-LBTWDOQPSA-N
SMILES
[2H]C1([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C1([2H])[2H]
Appearance (Color)
Clear colorless
Proton NMR
≤ 300ppm
Assay from Supplier's CofA
≥99.5% ¹H nmr
Form
Liquid
Assay from Supplier's CofA
≥99.5% Isotopic enrichment by ¹H nmr

Description

It is a common solvent used in NMR spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text