Thermo Scientific Chemicals

1,2-Dichloroethane-d{4}, 99% (Isotopic), Thermo Scientific Chemicals

Catalog number: 042281.03
1 g, Each
Thermo Scientific Chemicals

1,2-Dichloroethane-d{4}, 99% (Isotopic), Thermo Scientific Chemicals

Catalog number: 042281.03
1 g, Each
Quantity
Catalog number: 042281.03
also known as 042281-03
Price (USD)
Have Questions?

Chemical Identifiers

CAS
17060-07-0
IUPAC Name
1,2-dichloroethane
Molecular Formula
C2H4Cl2
InChI Key
WSLDOOZREJYCGB-UHFFFAOYSA-N
SMILES
ClCCCl
Form
Liquid
Assay from Supplier's CofA
≥99.0% Isotopic enrichment by ¹H nmr
Assay (GC)
≥99.0%
Assay from Supplier's CofA
≥99.0% ¹H nmr
Water Content (Karl Fischer Titration)
≤ 300ppm

Description

1,2-Dichloroethane-d4 is widely used as a solvent for NMR analysis, and in the synthesis of poly(ethylene-d4 oxide) as a chain termination reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text