N,N-Dimethylformamide-d{7}, 99.5% (Isotopic), Thermo Scientific Chemicals
N,N-Dimethylformamide-d{7}, 99.5% (Isotopic), Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N-Dimethylformamide-d{7}, 99.5% (Isotopic), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 x 1 g
1 g
5 g
10 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 042284.46
also known as 042284-46
Price (USD)/ Each
234.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
276.00 
Save 41.35 (15%)
-
Add to cart
Quantity:
5 x 1 g
Request bulk or custom format
Price (USD)/ Each
234.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
276.00 
Save 41.35 (15%)
Add to cart
N,N-Dimethylformamide-d{7}, 99.5% (Isotopic), Thermo Scientific Chemicals
Catalog number042284.46
Price (USD)/ Each
234.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
276.00 
Save 41.35 (15%)
-
Add to cart
Chemical Identifiers
CAS4472-41-7
IUPAC NameN,N-di(²H₃)methyl(²H)formamide
Molecular FormulaC3H7NO
InChI KeyZMXDDKWLCZADIW-YYWVXINBSA-N
SMILES[2H]C(=O)N(C([2H])([2H])[2H])C([2H])([2H])[2H]
View more
SpecificationsSpecification SheetSpecification Sheet
Residual water200 ppm max
Assay (unspecified)99.5% min isotopic enrichment
It is a common solvent used in NMR spectroscopy. It is also used as a reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only