Thermo Scientific Chemicals

Ethylene glycol-d{6}, 98% (Isotopic), Thermo Scientific Chemicals

Catalog number: 042288.03
1 g, Each
Thermo Scientific Chemicals

Ethylene glycol-d{6}, 98% (Isotopic), Thermo Scientific Chemicals

Catalog number: 042288.03
1 g, Each
Quantity
Catalog number: 042288.03
also known as 042288-03
Price (USD)
Have Questions?

Chemical Identifiers

CAS
15054-86-1
IUPAC Name
(²H₄)ethane-1,2-di(²H)ol
Molecular Formula
C2H6O2
InChI Key
LYCAIKOWRPUZTN-UFSLNRCZSA-N
SMILES
[2H]OC([2H])([2H])C([2H])([2H])O[2H]
Appearance (Color)
Clear colorless
Form
Liquid
Assay from Supplier's CofA
≥98.0% Isotopic enrichment by ¹H nmr
Assay from Supplier's CofA
≥98.0% ¹H nmr
GC/MS Analysis
≥98.0%

Description

It is used in synthetic materials and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text