Search Thermo Fisher Scientific
n-Heptane-d{16} is used as a test fuel component in anti-knock test engines, a 100% heptane fuel is the zero point of the octane rating scale. Heptane is commercially available as mixed isomers for use in paints and coatings, as the rubber cement solvent, n-heptane for research and development and pharmaceutical manufacturing and as a minor component of gasoline.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.