Thermo Scientific Chemicals

1,1,1,3,3,3-Hexafluoro-2-propanol-d{2}, 98%(Isotopic), Thermo Scientific Chemicals

Catalog number: 042291.03
1 g, Each
Thermo Scientific Chemicals

1,1,1,3,3,3-Hexafluoro-2-propanol-d{2}, 98%(Isotopic), Thermo Scientific Chemicals

Catalog number: 042291.03
1 g, Each
Quantity
Catalog number: 042291.03
also known as 042291-03
Price (USD)
Have Questions?

Chemical Identifiers

CAS
38701-74-5
IUPAC Name
hexafluoro(²H)propan-2-(²H)ol
Molecular Formula
C3H2F6O
InChI Key
BYEAHWXPCBROCE-AWPANEGFSA-N
SMILES
[2H]OC([2H])(C(F)(F)F)C(F)(F)F
GC/MS Analysis
≥98.0%
Purity
1H NMR For Chemical Purity: ≥98.0%
Purity
1H NMR for Isotopic Enrichment: ≥98.0%
Water Content
≤200 ppm

Description

1,1,1,3,3,3-Hexafluoro-2-propanol-d{2} is a stable isotope used for biochemical research. It is also used as an intermediate in chemical research and in pharmaceutics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text