Quartz rod, fused, 5.0mm (0.197in) dia, Thermo Scientific Chemicals
Quartz rod, fused, 5.0mm (0.197in) dia, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Quartz rod, fused, 5.0mm (0.197in) dia, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
48 in.
12 in.
Catalog number 042293.NT
also known as 042293-NT
Price (USD)/ Each
44.40
-
Add to cart
Quantity:
48 in.
Request bulk or custom format
Price (USD)/ Each
44.40
Add to cart
Quartz rod, fused, 5.0mm (0.197in) dia, Thermo Scientific Chemicals
Catalog number042293.NT
Price (USD)/ Each
44.40
-
Add to cart
Chemical Identifiers
CAS14808-60-7
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormRod, 5.0 mm (0.197 in.) dia.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only