Thermo Scientific Chemicals

2-Ethylhexyl methacrylate, 98%, stab. with 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: 042303.AK
250 mL, Each
Thermo Scientific Chemicals

2-Ethylhexyl methacrylate, 98%, stab. with 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: 042303.AK
250 mL, Each
Quantity
Catalog number: 042303.AK
also known as 042303-AK
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
688-84-6
IUPAC Name
2-ethylhexyl 2-methylprop-2-enoate
Molecular Formula
C12H22O2
InChI Key
WDQMWEYDKDCEHT-UHFFFAOYNA-N
SMILES
CCCCC(CC)COC(=O)C(C)=C
Color (APHA)
≤10
Stabilizer
40-60ppm MEHQ
Assay from Supplier's CofA
≥97.5%

Description

Monomer used in the preparation of polymethacrylate polymers

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text