Thermo Scientific Chemicals

Tri(1-naphthyl)phosphine, 97%, Thermo Scientific Chemicals

Catalog number: 042304.02
0.5 g, Each
Thermo Scientific Chemicals

Tri(1-naphthyl)phosphine, 97%, Thermo Scientific Chemicals

Catalog number: 042304.02
0.5 g, Each
Quantity
Catalog number: 042304.02
also known as 042304-02
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3411-48-1
IUPAC Name
tris(naphthalen-1-yl)phosphane
Molecular Formula
C30H21P
InChI Key
DMEUUKUNSVFYAA-UHFFFAOYSA-N
SMILES
C1=CC2=CC=CC(P(C3=CC=CC4=CC=CC=C34)C3=C4C=CC=CC4=CC=C3)=C2C=C1
Proton NMR
Conforms to structure
Appearance (Color)
White to pale yellow
Form
Powder
Phosphorus-31 NMR
Conforms to structure
Purity (DSC)
>96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text