Thermo Scientific Chemicals

Chromium carbide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 042313.09
10 g, Each
Thermo Scientific Chemicals

Chromium carbide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 042313.09
10 g, Each
Quantity
Catalog number: 042313.09
also known as 042313-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12105-81-6
Total Metal Impurities
0.5% max.

Description

Chromium carbide is useful in the surface treatment of metal components. Chromium carbide is used to coat the surface of another metal in a technique known as thermal spraying. Chromium carbide is used as an additive in cutting tools made out of cemented carbides, in order to improve toughness by preventing the growth of large grains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text