Thermo Scientific Chemicals

Methanol-d{4}, 99.8% (Isotopic), contains 0.05% v/v TMS, Thermo Scientific Chemicals

Catalog number: 042319.KV
10 Ea., Each
Thermo Scientific Chemicals

Methanol-d{4}, 99.8% (Isotopic), contains 0.05% v/v TMS, Thermo Scientific Chemicals

Catalog number: 042319.KV
10 Ea., Each
Quantity
Catalog number: 042319.KV
also known as 042319-KV
Price (USD)
Have Questions?

Chemical Identifiers

CAS
811-98-3
IUPAC Name
(²H₃)methan(²H)ol
Molecular Formula
CH4O
InChI Key
OKKJLVBELUTLKV-MZCSYVLQSA-N
SMILES
[2H]OC([2H])([2H])[2H]
Assay (GC)
≥ 99.5%
Form
Liquid
Assay from Supplier's CofA
conforms by ¹H nmr
Assay from Supplier's CofA
≥99.78% Isotopic enrichment by ¹H nmr
Water Content (Karl Fischer Titration)
≤ 300ppm

Description

Methyl alcohol-d4 is the labeled analog of methyl alcohol, a common laboratory solvent. It is a common solvent used in NMR spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text