Aluminum slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length, 99.99% (metals basis), Thermo Scientific Chemicals
Aluminum slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length, 99.99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 042328.18
also known as 042328-18
Price (USD)/ Each
62.60
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
62.60
Add to cart
Aluminum slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number042328.18
Price (USD)/ Each
62.60
-
Add to cart
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery white
FormSlug, 6.35mm (0.25in) diameter.
Assay from Supplier's CofA≥99.99% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only