Nickel slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Nickel slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Have Questions?
Quantity:
25 Slugs
Catalog number 042331.QK
also known as 042331-QK
Price (USD)/ Each
213.00
-
Add to cart
Quantity:
25 Slugs
Request bulk or custom format
Price (USD)/ Each
213.00
Add to cart
Nickel slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Catalog number042331.QK
Price (USD)/ Each
213.00
-
Add to cart
Chemical Identifiers
CAS7440-02-0
IUPAC Namenickel
Molecular FormulaNi
InChI KeyPXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery
Assay from Supplier's CofA≥99.995% (metals basis)
FormSlug, 6.35mm (0.25in) diameter.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only