Thermo Scientific Chemicals

Dichloromethane-d2, 100% (Isotopic), Thermo Scientific Chemicals

Catalog number: 042334.KV
10 Ea., Each
Thermo Scientific Chemicals

Dichloromethane-d2, 100% (Isotopic), Thermo Scientific Chemicals

Catalog number: 042334.KV
10 Ea., Each
Quantity
Catalog number: 042334.KV
also known as 042334-KV
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1665-00-5
IUPAC Name
dichloro(²H₂)methane
Molecular Formula
CH2Cl2
InChI Key
YMWUJEATGCHHMB-DICFDUPASA-N
SMILES
[2H]C([2H])(Cl)Cl
Form
Liquid
Assay from Supplier's CofA
≥99.95% Isotopic enrichment by ¹H nmr
Assay from Supplier's CofA
≥99.5% ¹H nmr
Water Content (Karl Fischer Titration)
≤ 300ppm
Appearance (Color)
Clear colorless

Description

Labeled methylene chloride is used in organic chemical reactions as a general solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text