Pyridine-d{5}, 99.5% (Isotopic), contains 0.05% v/v TMS, Thermo Scientific Chemicals
Pyridine-d{5}, 99.5% (Isotopic), contains 0.05% v/v TMS, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pyridine-d{5}, 99.5% (Isotopic), contains 0.05% v/v TMS, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 042342.06
also known as 042342-06
Price (USD)/ Each
101.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
119.00 
Save 17.35 (15%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
101.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
119.00 
Save 17.35 (15%)
Add to cart
Pyridine-d{5}, 99.5% (Isotopic), contains 0.05% v/v TMS, Thermo Scientific Chemicals
Catalog number042342.06
Price (USD)/ Each
101.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
119.00 
Save 17.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7291-22-7
IUPAC Name(²H₅)pyridine
Molecular FormulaC5H5N
InChI KeyJUJWROOIHBZHMG-RALIUCGRSA-N
SMILES[2H]C1=NC([2H])=C([2H])C([2H])=C1[2H]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥99.5%
FormLiquid
Assay from Supplier's CofA≥99.5% ¹H nmr
Assay from Supplier's CofA≥99.5% Isotopic enrichment by ¹H nmr
View more
Pyridine-d{5} is used in dehalogenation reactions and can be used as a base in condensation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only