Sodium deuteroxide, 40% w/w solution in D{2}O, 99.5%(Isotopic), Thermo Scientific Chemicals
Sodium deuteroxide, 40% w/w solution in D{2}O, 99.5%(Isotopic), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium deuteroxide, 40% w/w solution in D{2}O, 99.5%(Isotopic), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 042346.09
also known as 042346-09
Price (USD)/ Each
36.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
42.50 
Save 5.85 (14%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
36.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
42.50 
Save 5.85 (14%)
Add to cart
Sodium deuteroxide, 40% w/w solution in D{2}O, 99.5%(Isotopic), Thermo Scientific Chemicals
Catalog number042346.09
Price (USD)/ Each
36.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
42.50 
Save 5.85 (14%)
-
Add to cart
Chemical Identifiers
CAS14014-06-3
IUPAC Namesodium hydroxide
Molecular FormulaHNaO
InChI KeyHEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES[OH-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Isotopic purity99.5% min
Appearance (Color)Clear
FormLiquid
Assay (Titration)38.0 to 42.0 wt%
Sodium deuteroxide is a reagent used in the synthesis of rac Zearalenone-d6. is used in many chemical reactions and industrial practices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only