Thermo Scientific Chemicals

Sodium deuteroxide, 40% w/w solution in D{2}O, 99.5%(Isotopic), Thermo Scientific Chemicals

Catalog number: 042346.18
50 g, Each
Thermo Scientific Chemicals

Sodium deuteroxide, 40% w/w solution in D{2}O, 99.5%(Isotopic), Thermo Scientific Chemicals

Catalog number: 042346.18
50 g, Each
Quantity
Catalog number: 042346.18
also known as 042346-18
Price (USD)
Have Questions?

Chemical Identifiers

CAS
14014-06-3
IUPAC Name
sodium hydroxide
Molecular Formula
HNaO
InChI Key
HEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES
[OH-].[Na+]
Appearance (Color)
Clear
Form
Liquid
Isotopic purity
99.5% min
Assay (Titration)
38.0 to 42.0 wt%

Description

Sodium deuteroxide is a reagent used in the synthesis of rac Zearalenone-d6. is used in many chemical reactions and industrial practices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text