Thermo Scientific Chemicals

Toluene-d{8}, 100% (Isotopic), Thermo Scientific Chemicals

Catalog number: 042356.KV
10 Ea., Each
Thermo Scientific Chemicals

Toluene-d{8}, 100% (Isotopic), Thermo Scientific Chemicals

Catalog number: 042356.KV
10 Ea., Each
Quantity
Catalog number: 042356.KV
also known as 042356-KV
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2037-26-5
IUPAC Name
1-(²H₃)methyl(²H₅)benzene
Molecular Formula
C7H8
InChI Key
YXFVVABEGXRONW-JGUCLWPXSA-N
SMILES
[2H]C1=C([2H])C([2H])=C(C([2H])=C1[2H])C([2H])([2H])[2H]
Appearance (Color)
Colorless
Isotopic purity
≥99.4% (isotopic enrichment)
Transition Temperature
Liquid

Description

Toluene-d{8} is used to produce d7-4-methylacetophenone by heating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text