Trifluoroacetic acid-d, 99.5%(Isotopic), Thermo Scientific Chemicals
Trifluoroacetic acid-d, 99.5%(Isotopic), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trifluoroacetic acid-d, 99.5%(Isotopic), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 042357.09
also known as 042357-09
Price (USD)/ Each
44.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
52.60 
Save 7.95 (15%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
44.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
52.60 
Save 7.95 (15%)
Add to cart
Trifluoroacetic acid-d, 99.5%(Isotopic), Thermo Scientific Chemicals
Catalog number042357.09
Price (USD)/ Each
44.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
52.60 
Save 7.95 (15%)
-
Add to cart
Chemical Identifiers
CAS599-00-8
IUPAC Nametrifluoroethan(²H)oic acid
Molecular FormulaC2HF3O2
InChI KeyDTQVDTLACAAQTR-DYCDLGHISA-N
SMILES[2H]OC(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.5% ¹H nmr
Appearance (Color)Clear colorless
Assay from Supplier's CofA≥99.5% Isotopic enrichment by ¹H nmr
FormLiquid
Trifluoroacetic acid-d is employed as solvent to investigate the conformations of cyclic peptides by proton NMR spectra. It is also used to compose NMR solvent mixture to investigate the NMR spectra of high molecular weight polyesters such as polyglycolide (PG) and poly(ethylene terephthalate)and as a biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only